Learn More
Medchemexpress LLC TC-A 2317 hydrochloride | 1245907-03-2 | MFCD19690936 | 99.6% | 392.93 g/mol | C19H29ClN6O | 10 MG
SDP
Catalog No. 5000247082
Encompass_Preferred
TC-A 2317 hydrochloride is the hydrochloride salt of TC-A 2317, a potent, orally active Aurora A kinase inhibitor characterized for cellular activity and antitumor effects. It is supplied as a high-purity research reagent suitable for in vitro and in vivo pharmacology studies.
  • Potent Aurora A kinase inhibitor (Ki = 1.2 nM).
  • Selective over Aurora B kinase (Ki = 101 nM) and many other kinases.
  • Hydrochloride salt form for improved stability and solubility.
  • High purity suitable for research (≈99.6%).
  • Available in small milligram quantities for preclinical studies.
  • Molecular weight 392.93 g/mol; molecular formula C19H29ClN6O.
Catalog No. 50-002-47082 Supplier Medchemexpress LLC Supplier No. HY10326610MG
TC-A 2317 hydrochloride is the hydrochloride salt of TC-A 2317, a potent, orally active Aurora A kinase inhibitor characterized for cellular activity and antitumor effects. It is supplied as a high-purity research reagent suitable for in vitro and in vivo pharmacology studies.
  • Potent Aurora A kinase inhibitor (Ki = 1.2 nM).
  • Selective over Aurora B kinase (Ki = 101 nM) and many other kinases.
  • Hydrochloride salt form for improved stability and solubility.
  • High purity suitable for research (≈99.6%).
  • Available in small milligram quantities for preclinical studies.
  • Molecular weight 392.93 g/mol; molecular formula C19H29ClN6O.