Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ TC Adson Forceps - Brown, Dressing, Tissue

Catalog No. 21124576 Shop All Surgical Design Products
Change view
Click to view available options
Style:
Brown Tissue Forceps
Dressing
Tissue Forceps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
21-124-576 Dressing
21-124-564 Brown Tissue Forceps
21-124-570 Tissue Forceps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-124-576 Supplier Surgical Design™ Supplier No. 249TC
Add to Cart
Add to Cart

TC Adson forceps with tungsten carbide inserts deliver precision grip and lasting performance for delicate tissue handling.

  • Constructed from premium surgical-grade stainless steel with tungsten carbide (TC) inserts for enhanced grip, wear resistance, and control
  • Available in Dressing, Tissue, and Brown formats to accommodate a range of surgical applications
  • Fine, serrated or toothed tips ensure precise handling of tissue, sutures, and dressings
  • TC inserts provide non-slip holding power and exceptional edge longevity
  • Gold-plated handles distinguish TC instruments for easy identification
  • Ergonomic design allows secure handling with minimal hand fatigue during delicate work
  • Lightweight and balanced construction enhances tactile feedback and precision
  • Autoclavable and reusable, designed for professional surgical and laboratory use
  • Polished stainless finish for easy cleaning and long-term corrosion protection
  • Ideal for grasping tissue, holding sutures, and manipulating dressings
  • Commonly used in general surgery, plastic, dental, and microsurgical procedures
  • Suitable for clinical, veterinary, and training environments

Specifications

Product Type Thumb Forceps
Autoclavable Autoclavable
Certifications/Compliance ISO, GMP
Coating Tungsten Carbide
Color Multi-colored
Material Stainless Steel
Disposable Reusable
Latex Free Yes
Style Dressing
Teeth Count Serrated
Length (English) 4.75 in.
Length (Metric) 12 cm
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.