Learn More
Electron Microscopy Sciences Thermally Conductive Adhesive ETC Bond 568 25 GR
SDP
Supplier:  Electron Microscopy Sciences 50380568
Encompass_Preferred
Thermally conductive adhesive, Aluminum filled, 1:1 epoxy paste, Excellent mechanical strength to 400 deg F.
Catalog No. 50-997-384