Learn More
Electron Microscopy Sciences Thermally Conductive Adhesive ETC Bond 805 25 GR
SDP
Catalog No. 50997387
Encompass_Preferred
Thermally conductive adhesive, Aluminum filled, Two part epoxy paste, Low shrink rate, Excellent mechanical strength to 570 deg F.
Catalog No. 50-997-387 Supplier Electron Microscopy Sciences Supplier No. 50380805
Thermally conductive adhesive, Aluminum filled, Two part epoxy paste, Low shrink rate, Excellent mechanical strength to 570 deg F.