Learn More
Electron Microscopy Sciences Thermally Conductive Adhesive ETC Bond 860 25 GR
SDP
Catalog No. 50997388
Encompass_Preferred
Thermally conductive adhesive, Aluminum nitride filled, 1:1 epoxy paste, Excellent mechanical & thermal properties to 400 deg F.
Catalog No. 50-997-388 Supplier Electron Microscopy Sciences Supplier No. 50380860
Thermally conductive adhesive, Aluminum nitride filled, 1:1 epoxy paste, Excellent mechanical & thermal properties to 400 deg F.