Learn More
Accuflow Systems 3/4" Polypropylene End Caps, Pack of 10
Catalog No. 501108242
Encompass_Preferred
3/4" Polypropylene End Caps are designed for use in GMP Manufacturing and laboratories. These caps can be autoclavable as well as gama irradiated. Constructed of USP Class VI Polypropylene. Designed for Sanitary applications. Pack of 10
Catalog No. 50-110-8242 Supplier Accuflow Systems Supplier No. PPCAP075
3/4" Polypropylene End Caps are designed for use in GMP Manufacturing and laboratories. These caps can be autoclavable as well as gama irradiated. Constructed of USP Class VI Polypropylene. Designed for Sanitary applications. Pack of 10