Learn More
TARGETMOL CHEMICALS INC TC-DAPK 6 5MG
SDP
Catalog No. 502581131
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TC-DAPK 6 is an ATP-competitive inhibitor of Death-associated protein kinase (DAPK) with IC50s of 69 and 225 nM for DAPK1 and DAPK3. purity: 98%
Catalog No. 50-258-1131 Supplier TARGETMOL CHEMICALS INC Supplier No. T170045MG
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TC-DAPK 6 is an ATP-competitive inhibitor of Death-associated protein kinase (DAPK) with IC50s of 69 and 225 nM for DAPK1 and DAPK3. purity: 98%