Learn More
TARGETMOL CHEMICALS INC TC-E 5003 200MG
SDP
Catalog No. 502879140
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TC-E 5003 (NSC-30176) is a selective inhibitor of PRMT1 with IC50 of 1.5 uM. Purity 97.01%
Catalog No. 50-287-9140 Supplier TARGETMOL CHEMICALS INC Supplier No. T21826200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TC-E 5003 (NSC-30176) is a selective inhibitor of PRMT1 with IC50 of 1.5 uM. Purity 97.01%