Learn More
Medchemexpress LLC TC-G 1005 | 1415407-60-1 | 99.3% | 399.48 | 100 MG
SDP
Catalog No. 5000343799
Encompass_Preferred
TC-G 1005 is a potent, selective, and orally active agonist of the BA receptor Takeda G protein-coupled receptor 5 (TGR5), with EC50s of 0.72 nM for hTGR5 and 6.2 nM for mTGR5. It can reduce glucose levels in vivo.
  • Potent and selective TGR5 agonist.
  • Orally active.
  • Activates human and mouse TGR5.
  • Stimulates GLP-1 secretion.
  • Reduces blood glucose levels.
Catalog No. 50-003-43799 Supplier Medchemexpress LLC Supplier No. HY11017310MG
TC-G 1005 is a potent, selective, and orally active agonist of the BA receptor Takeda G protein-coupled receptor 5 (TGR5), with EC50s of 0.72 nM for hTGR5 and 6.2 nM for mTGR5. It can reduce glucose levels in vivo.
  • Potent and selective TGR5 agonist.
  • Orally active.
  • Activates human and mouse TGR5.
  • Stimulates GLP-1 secretion.
  • Reduces blood glucose levels.