Learn More
Medchemexpress LLC HY-110173 5mg Medchemexpress, TC-G 1005 CAS:1415407-60-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1101735MG
Encompass_Preferred
Medchemexpress, HY-110173 5mg TC-G 1005 CAS:1415407-60-1 TC-G 1005 is a potent, selective and orally active agonist of the BA receptor Takeda G protein-coupled receptor 5 (TGR5), with EC50s of 0.72 and 6.2 nM for hTGR5 and mTGR5, respectively. TC-G 1005 can reduce glucose levels in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0780