Learn More
TARGETMOL CHEMICALS INC TC-G-1008 10MG
SDP
Catalog No. 502579856
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TC-G-1008 (GPR39-C3) is a GPR39 (zinc receptor) agonist (EC50 values are 0.4 and 0.8 nM for rat and human receptors respectively). purity: 99%
Catalog No. 50-257-9856 Supplier TARGETMOL CHEMICALS INC Supplier No. T464810MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TC-G-1008 (GPR39-C3) is a GPR39 (zinc receptor) agonist (EC50 values are 0.4 and 0.8 nM for rat and human receptors respectively). purity: 99%