Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TC-G-1008 | 1621175-65-2 | 99.4% | 418.90 | C18H19ClN6O2S | 10 MG
SDP

Catalog No. 5000246932
Encompass_Preferred

TC-G-1008 (GPR39-C3) is a small-molecule, orally available agonist of the G protein-coupled receptor GPR39, supplied as a high-purity solid for in vitro and in vivo pharmacology. It exhibits sub-nanomolar potency (EC50 0.4 nM in rat, 0.8 nM in human) and is suitable for receptor pharmacology, target validation, and pharmacodynamic studies.

  • Molecular formula: C18H19ClN6O2S
  • Molecular weight: 418.90
  • Purity: 99.4%
  • Physical form: solid (powder)
  • Solubility: DMSO ≥100 mg/mL; in vivo formulations ≥2.5 mg/mL
  • Suggested storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years)
  • Available pack sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg

Catalog No. 50-002-46932 Supplier Medchemexpress LLC Supplier No. HY10300710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TC-G-1008 (GPR39-C3) is a small-molecule, orally available agonist of the G protein-coupled receptor GPR39, supplied as a high-purity solid for in vitro and in vivo pharmacology. It exhibits sub-nanomolar potency (EC50 0.4 nM in rat, 0.8 nM in human) and is suitable for receptor pharmacology, target validation, and pharmacodynamic studies.

  • Molecular formula: C18H19ClN6O2S
  • Molecular weight: 418.90
  • Purity: 99.4%
  • Physical form: solid (powder)
  • Solubility: DMSO ≥100 mg/mL; in vivo formulations ≥2.5 mg/mL
  • Suggested storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years)
  • Available pack sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.