Learn More
Medchemexpress LLC HY-103007 5mg Medchemexpress, TC-G-1008 CAS:1621175-65-2 Purity:>98%
SDP
Catalog No. 501968009
Encompass_Preferred
Medchemexpress, HY-103007 5mg TC-G-1008 CAS:1621175-65-2 TC-G-1008 (GPR39-C3) is a potent and orally available GPR39 agonist with EC50 values of 0.4 and 0.8 nM for rat and human receptors respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8009 Supplier Medchemexpress LLC Supplier No. HY1030075MG
Medchemexpress, HY-103007 5mg TC-G-1008 CAS:1621175-65-2 TC-G-1008 (GPR39-C3) is a potent and orally available GPR39 agonist with EC50 values of 0.4 and 0.8 nM for rat and human receptors respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.