Learn More
Selleck Chemical LLC TC-H 106
SDP
Supplier:  Selleck Chemical LLC S67385MG
Encompass_Preferred
TC-H 106 (Pimelic Diphenylamide 106) is a slow tight-binding inhibitor of class I histone deacetylases(HDAC) with Ki value of 148 nM about 102 nM 14 nM for HDAC1 HDAC2 HDAC3 respectively
Catalog No. 50-217-3217