Learn More
TARGETMOL CHEMICALS INC TC HSD 21 5MG
SDP
Catalog No. 502581382
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TC HSD 21 is a potent and highly selective inhibitor of 17(beta)-hydroxysteroid dehydrogenase type 3 (IC50 = 14 nM). purity: 98%
Catalog No. 50-258-1382 Supplier TARGETMOL CHEMICALS INC Supplier No. T131005MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TC HSD 21 is a potent and highly selective inhibitor of 17(beta)-hydroxysteroid dehydrogenase type 3 (IC50 = 14 nM). purity: 98%