Learn More
Medchemexpress LLC HY-103394 5mg Medchemexpress, TC HSD 21 CAS:330203-01-5 Purity:>98%
SDP
Catalog No. 501968096
Encompass_Preferred
Medchemexpress, HY-103394 5mg TC HSD 21 CAS:330203-01-5 TC HSD 21 is a potent 17β-hydroxysteroid dehydrogenase type 3 (17β-HSD3) inhibitor with an IC50 of 14 nM. TC HSD 21 shows excellent selectivity over 17β-HSD isoenzymes and nuclear receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8096 Supplier Medchemexpress LLC Supplier No. HY1033945MG
Medchemexpress, HY-103394 5mg TC HSD 21 CAS:330203-01-5 TC HSD 21 is a potent 17β-hydroxysteroid dehydrogenase type 3 (17β-HSD3) inhibitor with an IC50 of 14 nM. TC HSD 21 shows excellent selectivity over 17β-HSD isoenzymes and nuclear receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.