Learn More
Medchemexpress LLC TC-I 15 10mM 1mL | 916734-43-5 | 10MM 1ML
SDP
Catalog No. 5000240685
Encompass_Preferred
TC-I 15 (TC-I-15) is a type of allosteric collagen-binding integrin 2 1 inhibitor and it also inhibits 1 1 and 11 1 TC-I 15 inhibits platelet adhesion to collagen and thrombus deposition TC-I 15 prevents the formation of a pre-metastatic microenvironment by inhibiting the uptake of cancer-associated fibroblast (CAF) extracellular vesicles (EVs) by lung fibroblasts which reduces the metastasis of salivary gland adenocystic carcinoma (SACC) to the lungs in mouse models [1 [2
Catalog No. 50-002-40685 Supplier Medchemexpress LLC Supplier No. HY10758810MM 1ML
TC-I 15 (TC-I-15) is a type of allosteric collagen-binding integrin 2 1 inhibitor and it also inhibits 1 1 and 11 1 TC-I 15 inhibits platelet adhesion to collagen and thrombus deposition TC-I 15 prevents the formation of a pre-metastatic microenvironment by inhibiting the uptake of cancer-associated fibroblast (CAF) extracellular vesicles (EVs) by lung fibroblasts which reduces the metastasis of salivary gland adenocystic carcinoma (SACC) to the lungs in mouse models [1 [2