Learn More
TARGETMOL CHEMICALS INC TC-I 2014 5MG
SDP
Catalog No. 502581129
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. TC-I 2014 shows antiallodynic properties in pain models. TC-I 2014 is a potent and orally active Benzimidazole-containing transient receptor potential melastatin 8 antagonist (IC50 0.8 nM 3.0 nM and 4.4 nM for canine human and rat channels respectively). purity: 98%
Catalog No. 50-258-1129 Supplier TARGETMOL CHEMICALS INC Supplier No. T170065MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. TC-I 2014 shows antiallodynic properties in pain models. TC-I 2014 is a potent and orally active Benzimidazole-containing transient receptor potential melastatin 8 antagonist (IC50 0.8 nM 3.0 nM and 4.4 nM for canine human and rat channels respectively). purity: 98%