Learn More
Medchemexpress LLC HY-110199 5mg Medchemexpress, TC-I 2014 CAS:1221349-53-6 Purity:>98%
SDP
Catalog No. 501968769
Encompass_Preferred
Medchemexpress, HY-110199 5mg TC-I 2014 CAS:1221349-53-6 TC-I 2014 (compound 5) is a potent and orally active Benzimidazole-containing transient receptor potential melastatin 8 (TRPM8) antagonist, with IC50 values of 0.8 nM, 3.0 nM and 4.4 nM for canine, human and rat channels respectively. TC-I 2014 exhibits antiallodynic properties in pain models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8769 Supplier Medchemexpress LLC Supplier No. HY1101995MG
Medchemexpress, HY-110199 5mg TC-I 2014 CAS:1221349-53-6 TC-I 2014 (compound 5) is a potent and orally active Benzimidazole-containing transient receptor potential melastatin 8 (TRPM8) antagonist, with IC50 values of 0.8 nM, 3.0 nM and 4.4 nM for canine, human and rat channels respectively. TC-I 2014 exhibits antiallodynic properties in pain models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.