Learn More
Cole-Parmer Digi-Sense Type-J, Heavy Duty SS T-Handle Probe, 10" L, Mini-Connector, .250" Dia, Grounded
Catalog No. 501961605 Shop All Cole-Parmer Products
Encompass_Preferred
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C). Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Catalog No. 50-196-1605 Supplier Cole-Parmer Supplier No. 9360182
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C). Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.