Learn More
Cole-Parmer Digi-Sense Type-J, Kapton-Insulated Probe, Mini-Connector, Exposed, 5ft 30 -Gauge, 6 Pk
Catalog No. 501961500 Shop All Cole-Parmer Products
Encompass_Preferred
Exposed-junction probes are not recommended for use with aqueous solutions. Do not subject type J exposed-junction probes to moisture as they will rust.
Catalog No. 50-196-1500 Supplier Cole-Parmer Supplier No. 0850587
Exposed-junction probes are not recommended for use with aqueous solutions. Do not subject type J exposed-junction probes to moisture as they will rust.