Learn More
Cole-Parmer Digi-Sense Type J Kapton - Insulated Thermocouple Probe; 24 -Gauge; 10'L
Catalog No. 501961521 Shop All Cole-Parmer Products
Encompass_Preferred
Flexible Kapton-insulated probe is ideal for environments that require slightly higher temperatures than FEP/PFA can provide. All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J black. Response Time: 0.5 seconds or greater.
Catalog No. 50-196-1521 Supplier Cole-Parmer Supplier No. 0851790
Flexible Kapton-insulated probe is ideal for environments that require slightly higher temperatures than FEP/PFA can provide. All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J black. Response Time: 0.5 seconds or greater.