Learn More
Cole-Parmer Digi-Sense Type J Hermetically Sealed Tip Insulated Thermocouple, 10ft L, 24 Awg
Catalog No. 501961545 Shop All Cole-Parmer Products
Encompass_Preferred
Do not subject type J exposed-junction probes to moisture as they will rust
Catalog No. 50-196-1545 Supplier Cole-Parmer Supplier No. 1852525
Do not subject type J exposed-junction probes to moisture as they will rust