Learn More
Cole-Parmer Compact PFA-Coated Thermocouple Probe, 4.5" L, Grounded d; Type K
Catalog No. 501961492 Shop All Cole-Parmer Products
Encompass_Preferred
General-purpose probes are designed for use with liquids, gases and semisolids. The response time is 2 seconds or greater. Probe comes with PVC handle. color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue.
Catalog No. 50-196-1492 Supplier Cole-Parmer Supplier No. 0844112
General-purpose probes are designed for use with liquids, gases and semisolids. The response time is 2 seconds or greater. Probe comes with PVC handle. color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue.