Learn More
Cole-Parmer Digi-Sense Type-K, Extension Cable, Mini Connector, 25ft, 20-Gauge
Catalog No. 501961509 Shop All Cole-Parmer Products
Encompass_Preferred
This extension cable has a 20-gauge (0.032" dia) thermocouple wire with PVC/PVC insulation. Maximum temperature: 221 F (105 C).
Catalog No. 50-196-1509 Supplier Cole-Parmer Supplier No. 0851635
This extension cable has a 20-gauge (0.032" dia) thermocouple wire with PVC/PVC insulation. Maximum temperature: 221 F (105 C).