Learn More
Cole-Parmer Digi-Sense Type-K, FEP-Insulated Probe, Mini-Connector, Exposed, 10ft 24 -Gauge
Catalog No. 501961516 Shop All Cole-Parmer Products
Encompass_Preferred
FEP-Insulated probes provide quick readings in hard-to-reach areas. All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue. Response Time: 0.5 seconds or greater.
Catalog No. 50-196-1516 Supplier Cole-Parmer Supplier No. 0851682
FEP-Insulated probes provide quick readings in hard-to-reach areas. All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue. Response Time: 0.5 seconds or greater.