Learn More
Cole-Parmer Digi-Sense Type-K, Fiberglass-Insulated Probe, Mini-Connector, Exposed, 10ft 24 -Gauge
Catalog No. 501961506 Shop All Cole-Parmer Products
Encompass_Preferred
Probe features a flexible fiberglass insulated wire used for high-temperature measurements. All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue. Response Time: 0.5 seconds or greater.
Catalog No. 50-196-1506 Supplier Cole-Parmer Supplier No. 0851282
Probe features a flexible fiberglass insulated wire used for high-temperature measurements. All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue. Response Time: 0.5 seconds or greater.