Learn More
Cole-Parmer Digi-Sense Type-K, Hi-Temperature 12" L, .020" Dia Probe, Mini-Connector, Grounded , 3ft 24 -Gauge
Supplier:  Cole-Parmer 9363101
Encompass_Preferred
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C).
Catalog No. NC1474981