Learn More
Cole-Parmer Digi-Sense Type-K Pipe Plug Probe SS 1 / 4" NPT (M) Mini-Connector, 0.5" L Grounded 5ft Cable
Catalog No. 501961514 Shop All Cole-Parmer Products
Encompass_Preferred
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C)..
Catalog No. 50-196-1514 Supplier Cole-Parmer Supplier No. 0851674
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C)..