Learn More
Cole-Parmer Digi-Sense Type-K Hypodermic Probe Mini-Connector, 2.25" L .063 Dia Grounded 2.5ft FEP Cable
Catalog No. 501961501 Shop All Cole-Parmer Products
Encompass_Preferred
Hypodermic tip penetration probes are designed for soft and semi-soft materials. Ideal for checking food temperatures, the probe has a stainles steel handle and comes with color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue. Response Time: 0.5 -1.2 seconds or greater.
Catalog No. 50-196-1501 Supplier Cole-Parmer Supplier No. 0850592
Hypodermic tip penetration probes are designed for soft and semi-soft materials. Ideal for checking food temperatures, the probe has a stainles steel handle and comes with color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue. Response Time: 0.5 -1.2 seconds or greater.