Learn More
Cole-Parmer Digi-Sense Type-K Hvy-Dty Ind SS Handle Mini-Connector, 36" L .188 Dia Grounded 3ft Armor Cable
Catalog No. 501961515 Shop All Cole-Parmer Products
Encompass_Preferred
Flex armor is not water tight and is not recommended for damp or aqueous environments. Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C). Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Catalog No. 50-196-1515 Supplier Cole-Parmer Supplier No. 0851680
Flex armor is not water tight and is not recommended for damp or aqueous environments. Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C). Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.