Learn More
Medchemexpress LLC TC LPA5 4 | 1393814-38-4 | 99.7% | 410.89 | C23H23ClN2O3 | 50 MG
SDP
Catalog No. 5000242021
Encompass_Preferred
TC LPA5 4 is a selective, non-lipid antagonist of the lysophosphatidic acid receptor LPA5 used in research to probe LPA5-mediated signaling. It exhibits antagonist activity at human LPA5 (cellular IC50 ~0.8 μM), inhibits LPA-induced platelet aggregation, reduces proliferation and migration of thyroid cancer cells in vitro, and has shown efficacy in mouse xenograft studies.
Catalog No. 50-002-42021 Supplier Medchemexpress LLC Supplier No. HY10761550MG
Add to Cart
Add to Cart
TC LPA5 4 is a selective, non-lipid antagonist of the lysophosphatidic acid receptor LPA5 used in research to probe LPA5-mediated signaling. It exhibits antagonist activity at human LPA5 (cellular IC50 ~0.8 μM), inhibits LPA-induced platelet aggregation, reduces proliferation and migration of thyroid cancer cells in vitro, and has shown efficacy in mouse xenograft studies.