Learn More
Medchemexpress LLC TC LPA5 4 5mg | 1393814-38-4 | 5MG
SDP
Catalog No. 5000240252
Encompass_Preferred
TC LPA5 4 is a LPA5 (GPR92)-specific non-lipid antagonist TC LPA5 4 inhibits LPA-induced aggregation of isolated human platelet (LPA5-RH7777 cell line) with an IC50 of 800 nM TC LPA5 4 displays selectivity for LPA5 over 80 other screened agent targets[1 TC LPA5 4 inhibits cell proliferation and migration of thyroid cancer cells[2
Catalog No. 50-002-40252 Supplier Medchemexpress LLC Supplier No. HY1076155MG
TC LPA5 4 is a LPA5 (GPR92)-specific non-lipid antagonist TC LPA5 4 inhibits LPA-induced aggregation of isolated human platelet (LPA5-RH7777 cell line) with an IC50 of 800 nM TC LPA5 4 displays selectivity for LPA5 over 80 other screened agent targets[1 TC LPA5 4 inhibits cell proliferation and migration of thyroid cancer cells[2