Learn More
Surgical Design™ TC Mayo Hegar Needle Holder Driver
Catalog No. 21124548 Shop All Surgical Design Products
Change view
Click to view available options
Length (English):
5 in.
5.5 in.
6 in.
7 in.
8 in.
10 in.
12 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
21-124-548 8 in.
21-124-541 5 in.
21-124-542 5.5 in.
21-124-544 6 in.
21-124-545 7 in.
21-124-546 10 in.
21-124-547 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 21-124-548 Supplier Surgical Design™ Supplier No. 202TC
Add to Cart
Add to Cart
TC Mayo-Hegar needle holder provides control and strength for suturing with long-lasting precision.
  • Made from high-quality surgical-grade stainless steel with tungsten carbide (TC) inserts for exceptional grip and durability
  • Available in 7 lengths — 5 in., 5.5 in., 6 in., 7 in., 8 in., 10 in., and 12 in. — to accommodate various needs
  • Cross-serrated jaws ensure a secure, non-slip hold on needles during suturing
  • TC inserts offer increased holding precision, maintaining sharpness and strength over extended use
  • Gold-plated handles distinguish TC instruments for quick identification
  • Ergonomic ring handle design provides excellent balance and control
  • Precision ratchet locking mechanism allows for adjustable tension and reliable hold
  • Autoclavable and reusable, built for heavy-duty hospital and surgical environments
  • Polished stainless finish for easy cleaning and long-term corrosion resistance
  • Ideal for suturing, wound closure, and general surgical procedures
  • Commonly used in medical, dental, veterinary, and autopsy applications
  • Excellent for operating rooms, training labs, and field surgical kits requiring precision and strength
Specifications
Product Type Needle Holder Driver
Autoclavable Autoclavable
Color Silver
Material Stainless Steel
Latex Free Yes
Style Cross-Serrated Jaw With Tungsten Carbide
Length (English) 8 in.
Length (Metric) 20.3 cm
Sterility Non-sterile