Learn More
Medchemexpress LLC HY-107623 1mg Medchemexpress, TC-MCH 7c CAS:864756-35-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1076231MG
Encompass_Preferred
Medchemexpress, HY-107623 1mg TC-MCH 7c CAS:864756-35-4 TC-MCH 7c, a phenylpyridone derivative, is an orally available, selective and brain-penetrable MCH1R antagonist with an IC50 of 5.6 nM for hMCH1R. TC-MCH 7c has Kis of 3.4 nM and 3.0 nM of human and mouse MCH1R, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8440