Learn More
Medchemexpress LLC HY-110115 10mg Medchemexpress, TC-Mps1-12 CAS:1206170-62-8 Purity:>98%
SDP
Catalog No. 501968748
Encompass_Preferred
Medchemexpress, HY-110115 10mg TC-Mps1-12 CAS:1206170-62-8 TC-Mps1-12 is a potent and selective monopolar spindle 1 (Mps1) inhibitor, with an IC50 of 6.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8748 Supplier Medchemexpress LLC Supplier No. HY11011510MG
Medchemexpress, HY-110115 10mg TC-Mps1-12 CAS:1206170-62-8 TC-Mps1-12 is a potent and selective monopolar spindle 1 (Mps1) inhibitor, with an IC50 of 6.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.