Learn More
Medchemexpress LLC HY-110115 5mg Medchemexpress, TC-Mps1-12 CAS:1206170-62-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1101155MG
Encompass_Preferred
Medchemexpress, HY-110115 5mg TC-Mps1-12 CAS:1206170-62-8 TC-Mps1-12 is a potent and selective monopolar spindle 1 (Mps1) inhibitor, with an IC50 of 6.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8749