Learn More
Bellco Glass, Inc. TC Nose Retrofit Kit With Four Filters
SDP
Catalog No. 501944628
Encompass_Preferred
The tissue culture nose-piece eliminates the possibility of cross-contamination due to over-pipetting. It incorporates a dual hydrophilic 0.8 ?m, hydrophobic 0.8 ?m filter which prevents further use once contaminated by fluid.
Catalog No. 50-194-4628 Supplier Bellco Glass, Inc. Supplier No. 122588050
The tissue culture nose-piece eliminates the possibility of cross-contamination due to over-pipetting. It incorporates a dual hydrophilic 0.8 ?m, hydrophobic 0.8 ?m filter which prevents further use once contaminated by fluid.