Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TC-O 9311, Tocris Bioscience™

Catalog No. 425510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-551-0 10 mg
42-555-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-551-0 Supplier R&D Systems Supplier No. 4255/10
This item is not returnable. View return policy

Potent and selective GPR139 agonist

Potent GPR139 agonist (EC50 = 39 nM in CHO-K1 cells expressing human GPR139). Displays no activity against a range of 90 diverse targets.

Chemical Identifiers

CAS 444932-31-4
Molecular Formula C20H19N3O4
Molecular Weight (g/mol) 365.39
MDL Number MFCD02372953
InChI Key KPTMSQHTGZMEFU-UHFFFAOYSA-N
Synonym 2-3,5-dimethoxybenzoyl-n-1-naphthyl hydrazinecarboxamide, 1-3,5-dimethoxybenzoyl amino-3-naphthalen-1-ylurea, n'-1-naphthyl amino carbonyl-3,5-dimethoxybenzhydrazide, 2-3,5-dimethoxyphenyl carbonyl-n-naphthalen-1-yl hydrazinecarboxamide, 3,5-dimethoxybenzoic acid 2-1-naphthalenylamino carbonyl hydrazide
PubChem CID 3376937
IUPAC Name 3,5-dimethoxy-N-{[(naphthalen-1-yl)carbamoyl]amino}benzamide
SMILES COC1=CC(=CC(OC)=C1)C(=O)NNC(=O)NC1=C2C=CC=CC2=CC=C1

Specifications

Quantity 10 mg
Formula Weight 365.38
Percent Purity >98%
Chemical Name or Material TC-O 9311
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.