Learn More
Sarstedt Inc Cell culture plate, 12 well, surface: Standard, flat base, 50/case
Catalog No. NC1408617 Shop All Sarstedt Inc. Products
Encompass_Preferred
12-well cell culture plate has a standard TC surface for adherent cells. TC Tested (sterile, non-pyrogenic/endotoxin-free, non-cytotoxic, DNase-/RNase-/DNA-free)
Catalog No. NC1408617 Supplier Sarstedt Inc Supplier No. 83.3921
12-well cell culture plate has a standard TC surface for adherent cells. TC Tested (sterile, non-pyrogenic/endotoxin-free, non-cytotoxic, DNase-/RNase-/DNA-free)