Learn More
Sarstedt Inc Cell culture plate, 48 well, surface: Cell+, flat base
Catalog No. NC1787625 Shop All Sarstedt Inc. Products
Encompass_Preferred
Cell culture plate, 48-well, PS, surface: Cell+ for sensitive adherent cells (e.g. primary cells), yellow, flat base, TC Tested (sterile, non-pyrogenic/endotoxin-free, non-cytotoxic, DNase-/RNase-/DNA-free).
Catalog No. NC1787625 Supplier Sarstedt Inc Supplier No. 83.3923.300
Cell culture plate, 48-well, PS, surface: Cell+ for sensitive adherent cells (e.g. primary cells), yellow, flat base, TC Tested (sterile, non-pyrogenic/endotoxin-free, non-cytotoxic, DNase-/RNase-/DNA-free).