Learn More
TARGETMOL CHEMICALS INC TC-S 7005 5MG
SDP
Catalog No. 502581128
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TC-S 7005 is an effective inhibitor of Polo-like kinases (IC50s 214 nM 4 nM and 24 nM for Plk1 Plk2 and Plk3). purity: 99%
Catalog No. 50-258-1128 Supplier TARGETMOL CHEMICALS INC Supplier No. T170085MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TC-S 7005 is an effective inhibitor of Polo-like kinases (IC50s 214 nM 4 nM and 24 nM for Plk1 Plk2 and Plk3). purity: 99%