Learn More
Medchemexpress LLC HY-108597 5mg Medchemexpress, TC-S 7005 CAS:1082739-92-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1085975MG
Encompass_Preferred
Medchemexpress, HY-108597 5mg TC-S 7005 CAS:1082739-92-1 TC-S 7005 is a Polo-like kinases (Plks) inhibitor with IC 50 s of 4 nM, 24 nM and 214 nM for Plk2, Plk3, and Plk1, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2323