Learn More
Cole-Parmer Digi-Sense Coiled Extension Cable, Type T, Male to Female Mini-Connector, 5 ft Length
Catalog No. NC2152099 Shop All Cole-Parmer Products
Encompass_Preferred
Each probe comes with color coded ANSI standard connectors: type J-black, type K-yellow, and type T-blue. Custom designs or changes to this product are available upon request.
This item is not returnable. View return policy
Catalog No. NC2152099 Supplier Cole-Parmer Supplier No. 9378504
Each probe comes with color coded ANSI standard connectors: type J-black, type K-yellow, and type T-blue. Custom designs or changes to this product are available upon request.