Learn More
Cole-Parmer Digi-Sense Type-T, Coiled Ext Cable, Male Mini Conn to Male Mini Conn, 5ft L
Catalog No. 501961610 Shop All Cole-Parmer Products
Encompass_Preferred
Thermocouple coiled extension cables for remote applications. Each probe comes with color coded ANSI standard connectors: type J-black, type K-yellow, and type T-blue. Maximum temperature: 221 F (105 C).
Catalog No. 50-196-1610 Supplier Cole-Parmer Supplier No. 9378514
Thermocouple coiled extension cables for remote applications. Each probe comes with color coded ANSI standard connectors: type J-black, type K-yellow, and type T-blue. Maximum temperature: 221 F (105 C).