Learn More
Cole-Parmer Digi-Sense Type-T, FEP-Insulated Probe, Mini-Connector, Exposed, 10ft 24 -Gauge
Catalog No. 501961517 Shop All Cole-Parmer Products
Encompass_Preferred
Exposed-junction probes are not recommended for use with aqueous solutions. Do not subject type J exposed-junction probes to moisture as they will rust
Catalog No. 50-196-1517 Supplier Cole-Parmer Supplier No. 0851683
Exposed-junction probes are not recommended for use with aqueous solutions. Do not subject type J exposed-junction probes to moisture as they will rust