Learn More
Cole-Parmer Digi-Sense Type-T, Fiberglass-Insulated Probe, Mini-Connector, Exposed, 10ft 24 -Gauge
Catalog No. 501961507 Shop All Cole-Parmer Products
Encompass_Preferred
Fiberglass wire is not recommended for use in moist or wet environments. Exposed-junction probes are not recommended for use with aqueous solutions. Do not subject type J exposed-junction probes to moisture as they will rust
Catalog No. 50-196-1507 Supplier Cole-Parmer Supplier No. 0851283
Fiberglass wire is not recommended for use in moist or wet environments. Exposed-junction probes are not recommended for use with aqueous solutions. Do not subject type J exposed-junction probes to moisture as they will rust