Learn More
Cole-Parmer Digi-Sense Type-T, Kapton-Insulated Probe, Mini-Connector, Exposed, 5ft 30 -Gauge, 6 Pk
Catalog No. 501961499 Shop All Cole-Parmer Products
Encompass_Preferred
Exposed-junction probes are not recommended for use with aqueous solutions. Do not subject type J exposed-junction probes to moisture as they will rust.
Catalog No. 50-196-1499 Supplier Cole-Parmer Supplier No. 0850585
Exposed-junction probes are not recommended for use with aqueous solutions. Do not subject type J exposed-junction probes to moisture as they will rust.