Learn More
Cole-Parmer Digi-Sense Type T Hermetically Sealed Tip Insulated Thermocouple, 20ft L, 24 Awg
Catalog No. 501961546 Shop All Cole-Parmer Products
Encompass_Preferred
Do not subject type J exposed-junction probes to moisture as they will rust
Catalog No. 50-196-1546 Supplier Cole-Parmer Supplier No. 1852532
Do not subject type J exposed-junction probes to moisture as they will rust