Learn More
Genesee Scientific Corporation Genesee Scientific GenClone Tissue Culture Flask, 250 ml, Sterile Gamma Irradiated, RNase/DNase-Free
SDP
Catalog No. NC2076248
Encompass
The Genesee Scientific GenClone Tissue Culture Flask is designed for optimal adherent cell culture applications. With a 250 ml capacity and striation-free, flat base, these flasks ensure reliable cell growth and enhance microscopy accuracy. Key features include a vented filter cap for sterile gas exchange and gamma irradiation for sterility assurance, making them a trusted choice for research and development in cell culture.
Catalog No. NC2076248 Supplier Genesee Scientific Corporation Supplier No. 25209
The Genesee Scientific GenClone Tissue Culture Flask is designed for optimal adherent cell culture applications. With a 250 ml capacity and striation-free, flat base, these flasks ensure reliable cell growth and enhance microscopy accuracy. Key features include a vented filter cap for sterile gas exchange and gamma irradiation for sterility assurance, making them a trusted choice for research and development in cell culture.